Sterigmatin ≥74%

Lieferant: Apollo Scientific
Achtung

BIBR1095-1MG
APOSBIBR1095-1MGEA 386 EUR
APOSBIBR1095-1MG
Sterigmatin ≥74%
Sterigmatin
Formel: C₁₇H₁₀O₆
Lagertemperatur: Kühlschrank
MDL: MFCD01725644
CAS-Nummer: 55256-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR