Vineomycin B2

Lieferant: Apollo Scientific
Achtung

BIBR1112-1MG
APOSBIBR1112-1MGEA 168 EUR
APOSBIBR1112-1MG
Vineomycin B2
Vineomycin B2
Formel: C₄₉H₅₈O₁₈
Lagertemperatur: Kühlschrank
MDL: MFCD09752773
CAS-Nummer: 80928-52-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR