Malayamycin A

Lieferant: Apollo Scientific
Achtung

BIBR1131-1MG
APOSBIBR1131-1MGEA 580 EUR
APOSBIBR1131-1MG
Malayamycin A
Malayamycin A
Formel: C₁₄H₂₀N₄O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD08457936
CAS-Nummer: 828914-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR