Sclerotiorin
Lieferant: Apollo Scientific
BIBR1146-5MG
APOSBIBR1146-5MGEA
1900
EUR
APOSBIBR1146-5MG
Sclerotiorin
Sclerotiorin
From microbial source
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
549-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...