Cefotetan

Lieferant: Apollo Scientific
Achtung

BIC0104-100MG BIC0104-250MG BIC0104-500MG
APOSBIC0104-100MGEA 21.9 EUR
APOSBIC0104-100MG APOSBIC0104-250MG APOSBIC0104-500MG
Cefotetan
Cefotetan
Formel: C₁₇H₁₇N₇O₈S₄
Lagertemperatur: Gefrierschrank
MDL: MFCD00864983
CAS-Nummer: 69712-56-7
EINECS: 274-093-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR