Carbenicillin Dinatriumsalz

Lieferant: Apollo Scientific
Gefahr

Synonyme: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

BIC0109-25G BIC0109-5G BIC0109-100G
APOSBIC0109-25GEA 161 EUR
APOSBIC0109-25G APOSBIC0109-5G APOSBIC0109-100G
Carbenicillin Dinatriumsalz
Carbenicillin Dinatriumsalz

Carbenicillin is an inhibitor of bacterial cell wall synthesisCell culture tested

Formel: C₁₇H₁₆N₂O₆S·2Na
Molecular Weight: 422.363 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00077683
CAS-Nummer: 4800-94-6
EINECS: 225-360-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR