Chloramphenicol ≥99%

Lieferant: Apollo Scientific
Gefahr

BIC0113-100G BIC0113-250G
APOSBIC0113-100GEA 190 EUR
APOSBIC0113-100G APOSBIC0113-250G
Chloramphenicol ≥99%
Chloramphenicol

Bacteriostatic antibiotic with activity against gram-positive and gram-negative bacteria.


  • Cell culture tested

Formel: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Schmelzpunkt: 148…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00078159
CAS-Nummer: 56-75-7
EINECS: 200-287-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR