Clindamycin hydrochloride

Lieferant: Apollo Scientific
Achtung

BIC0117-1G BIC0117-250MG
APOSBIC0117-1GEA 183 EUR
APOSBIC0117-1G APOSBIC0117-250MG
Clindamycin hydrochloride
Clindamycin hydrochloride

Clindamycin is a lincosamide antibiotic with a primarily bacteriostatic action against gram-positive bacteria.

Formel: C₁₈H₃₃ClN₂O₅S·HCl
Lagertemperatur: Kühlschrank
MDL: MFCD07793327
CAS-Nummer: 21462-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR