Folinic acid calcium salt ≥97,5%

Lieferant: Apollo Scientific
Achtung

BIC0607-1G BIC0607-5G BIC0607-25G
APOSBIC0607-1GEA 45.1 EUR
APOSBIC0607-1G APOSBIC0607-5G APOSBIC0607-25G
Folinic acid calcium salt ≥97,5%
Folinic acid calcium salt
Formel: C₂₀H₂₁CaN₇O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00006704
CAS-Nummer: 1492-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR