L(+)-Cystein Hydrochlorid

Lieferant: Apollo Scientific
Achtung

BIC0707-1KG BIC0707-500G
APOSBIC0707-1KGEA 0 EUR
APOSBIC0707-1KG APOSBIC0707-500G
L(+)-Cystein Hydrochlorid
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
EINECS: 200-157-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR