Cerulenin
Lieferant: Apollo Scientific
|
Achtung
|
BIC1007-5MG
APOSBIC1007-5MGEA
106
EUR
APOSBIC1007-5MG
Cerulenin
Cerulenin
Formel:
C₁₂H₁₇NO₃ Molecular Weight: 223.272 g/mol Schmelzpunkt: 93 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077686 CAS-Nummer: 17397-89-6 EINECS: 241-424-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...