Cytochalasin D

Lieferant: Apollo Scientific
Gefahr

Synonyme: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-trien-1,17-dion-21(R)-acetat

BIC1016-1MG BIC1016-5MG
APOSBIC1016-1MGEA 219 EUR
APOSBIC1016-1MG APOSBIC1016-5MG
Cytochalasin D
Cytochalasin D
Formel: C₃₀H₃₇NO₆
Molecular Weight: 507.63 g/mol
Schmelzpunkt: 260…267 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00077706
CAS-Nummer: 22144-77-0
EINECS: 244-804-1
UN: 1544
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR