Calyculin A

Lieferant: Apollo Scientific
Gefahr

BIC1019-10UG
APOSBIC1019-10UGEA 125 EUR
APOSBIC1019-10UG
Calyculin A
Calyculin A

PP1 inhibitor, PP2 inhibitor, serine/threonine phosphatatase inhibitor.

Formel: C₅₀H₈₁N₄O₁₅P
Lagertemperatur: Gefrierschrank
CAS-Nummer: 101932-71-2
UN: 3462
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR