3'-Desoxyadenosin (Cordycepin)

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3'-Deoxyadenosine, (2R,3R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol, Cordycepin

BIC1025-500MG
APOSBIC1025-500MGEA 132 EUR
APOSBIC1025-500MG
3'-Desoxyadenosin (Cordycepin)
3'-Desoxyadenosin (Cordycepin)
Formel: C₁₀H₁₃N₅O₃
Schmelzpunkt: 225 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00037998
CAS-Nummer: 73-03-0
UN: 3462
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR