Cefoperazone sodium salt

Lieferant: Apollo Scientific
Gefahr

BIC1033-25G BIC1033-5G
APOSBIC1033-25GEA 0 EUR
APOSBIC1033-25G APOSBIC1033-5G
Cefoperazone sodium salt
Cefoperazone sodium salt
Formel: C₂₅H₂₆N₉NaO₈S₂
Lagertemperatur: Kühlschrank
MDL: MFCD07793331
CAS-Nummer: 62893-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR