5(6)-Carboxyfluorescein diacetate
Lieferant: Apollo Scientific
Synonyme:
CFDA, 5(6)-Carboxy-di-O-acetylfluorescein, 5(6)-CFDA
BIC1066-100MG
BIC1066-250MG
APOSBIC1066-100MGEA
284
EUR
APOSBIC1066-100MG
APOSBIC1066-250MG
5(6)-Carboxyfluorescein diacetate
5(6)-Carboxyfluorescein diacetate
Formel:
C₂₅H₁₆O₉ Molecular Weight: 460.39 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467494 CAS-Nummer: 124387-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...