Curcumin 98%

Lieferant: Apollo Scientific
Achtung

BIC8500-5G BIC8500-100G BIC8500-25G
APOSBIC8500-5GEA 26.1 EUR
APOSBIC8500-5G APOSBIC8500-100G APOSBIC8500-25G
Curcumin 98%
Curcumin
Formel: C₂₁H₂₀O₆
Schmelzpunkt: 175 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00008365
CAS-Nummer: 458-37-7
EINECS: 207-280-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR