DSS (Disuccinimidylsuberat)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
DSS
BICL100-1G
APOSBICL100-1GEA
58
EUR
APOSBICL100-1G
DSS (Disuccinimidylsuberat)
DSS (Disuccinimidylsuberat)
Primary amine reactive -Receptor ligand cross-linking.
Formel:
C₁₆H₂₀N₂O₈ Molecular Weight: 368,34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049059 CAS-Nummer: 68528-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...