BS3 (Bis(sulphosuccinimidyl) suberate sodium salt)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
BS3
BICL101-100MG
APOSBICL101-100MGEA
306
EUR
APOSBICL101-100MG
BS3 (Bis(sulphosuccinimidyl) suberate sodium salt)
BS3 (Bis(sulphosuccinimidyl) suberate sodium salt)
Primary amine reactive -Membrane impermeableWater soluble.
Formel:
C₁₆H₁₈N₂Na₂O₁₄S₂ Molecular Weight: 572,43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054982 CAS-Nummer: 82436-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...