Bis[2-(sulphosuccinimidooxycarbonyloxy)ethyl]sulphone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sulpho BSOCOES
BICL105-100MG
APOSBICL105-100MGEA
116
EUR
APOSBICL105-100MG
Bis[2-(sulphosuccinimidooxycarbonyloxy)ethyl]sulphone
Bis[2-(sulphosuccinimidooxycarbonyloxy)ethyl]sulphone
Primary amine reactive -Base Cleavable -Water soluble
Ref: Zarling, D.A.Watson, A.and Bach, F.H.(1980) J.Immunol.124, 913-920
Formel:
C₁₄H₁₆N₂O₁₈S₃ Molecular Weight: 596,474 g/mol Schmelzpunkt: 169…173 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD07189459 CAS-Nummer: 57683-72-4 EINECS: 260-904-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...