Ethylene glycolbis(sulphosuccinimidylsuccinate)
Lieferant: Apollo Scientific
Achtung
Synonyme: Sulpho EGS
BICL109-100MG
APOSBICL109-100MGEA 267 EUR
APOSBICL109-100MG
Ethylene glycolbis(sulphosuccinimidylsuccinate)
Ethylene glycolbis(sulphosuccinimidylsuccinate)
Primary amine reactive -Hydroxylamine CleavableWater soluble
Ref: Abdella,R.M, Smith,P.K and Royer,G.P.(1979)Biochem.Biophys.Res.Commun.87, 734-742