Ethylene glycolbis(sulphosuccinimidylsuccinate)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sulpho EGS
BICL109-100MG
APOSBICL109-100MGEA
267
EUR
APOSBICL109-100MG
Ethylene glycolbis(sulphosuccinimidylsuccinate)
Ethylene glycolbis(sulphosuccinimidylsuccinate)
Primary amine reactive -Hydroxylamine CleavableWater soluble
Ref: Abdella,R.M, Smith,P.K and Royer,G.P.(1979)Biochem.Biophys.Res.Commun.87, 734-742