DPDPB (1,2-Di[3'-(2'-pyridyldithio)propionamido]butane)

Lieferant: Apollo Scientific
Achtung

Synonyme: DPDPB

BICL112-100MG
APOSBICL112-100MGEA 128 EUR
APOSBICL112-100MG
DPDPB (1,2-Di[3'-(2'-pyridyldithio)propionamido]butane)
DPDPB (1,2-Di[3'-(2'-pyridyldithio)propionamido]butane)

Sulphhydryl reactive -Cleavable

Formel: C₂₀H₂₆N₄O₂S₄
Molecular Weight: 482,706 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00153604
CAS-Nummer: 141647-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR