SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)

Lieferant: Apollo Scientific
Achtung

Synonyme: SMPB

BICL200-100MG
APOSBICL200-100MGEA 319 EUR
APOSBICL200-100MG
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)

Primary amine and Sulphhydryl reactive.Enzyme antibody conjugation - Spacer arm.

Formel: C₁₈H₁₆N₂O₆
Molecular Weight: 356,33 g/mol
Schmelzpunkt: 131 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00054961
CAS-Nummer: 79886-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR