Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)

Lieferant: Apollo Scientific
Achtung

BICL201-100MG
APOSBICL201-100MGEA 0 EUR
APOSBICL201-100MG
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)

Primary amine and Sulphhydryl reactiveSpacer arm -Water soluble


Ref: Bangs, J.D.et.al. (1986) J.Cell.Biol. 103, 255-263

Formel: C₁₈H₁₅N₂O₉S·Na
Lagertemperatur: Raumtemperatur
MDL: MFCD00054981
CAS-Nummer: 92921-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR