MBS (N-Succinimidyl 3-maleimidobenzoate)

Lieferant: Apollo Scientific
Achtung

Synonyme: MBS

BICL202-100MG BICL202-250MG
APOSBICL202-100MGEA 21.9 EUR
APOSBICL202-100MG APOSBICL202-250MG
MBS (N-Succinimidyl 3-maleimidobenzoate)
MBS (N-Succinimidyl 3-maleimidobenzoate)

Primary amine reactive -Sulphhydryl reactive. Protein conjugation Hapten carrier Protein conjugation

Formel: C₁₅H₁₀N₂O₆
Molecular Weight: 314.25 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005514
CAS-Nummer: 58626-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR