Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Lieferant: Apollo Scientific
Achtung

Synonyme: Sulfo MBS

BICL203-100MG
APOSBICL203-100MGEA 589 EUR
APOSBICL203-100MG
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Primary amine and sulphhydryl reactive -Water soluble


Ref: Aithal,H.N,et.al (1988) J.Immunol.Meth.112, 67-70

Formel: C₁₅H₉N₂O₉S·Na
Molecular Weight: 416,295 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01318594
CAS-Nummer: 92921-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR