Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sulfo SIAB
BICL205-100MG
APOSBICL205-100MGEA
270
EUR
APOSBICL205-100MG
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Primary amine and sulphhydryl reactive -Water soluble
Ref: Weltman, J.K. et.al. (1983) Bio.Techniques 1,148-152