Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)

Lieferant: Apollo Scientific
Achtung

Synonyme: Sulfo LC SPDP

BICL209-100MG
APOSBICL209-100MGEA 364 EUR
APOSBICL209-100MG
Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)
Sulpho-LC-SPDP (Sulphosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)

Water soluble -Spacer arm-SPDPantibody-toxin conjugationRef: Carlsson,J. et al (1978) Biochem.J. 173, 723-737

Formel: C₁₈H₂₂N₃NaO₈S₃
Molecular Weight: 527,567 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00083162
CAS-Nummer: 169751-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR