N-(E-Maleimidocaproyloxy)sulphosuccinimide

Lieferant: Apollo Scientific
Achtung

Synonyme: Sulpho EMCS

BICL221-50MG
APOSBICL221-50MGEA 139 EUR
APOSBICL221-50MG
N-(E-Maleimidocaproyloxy)sulphosuccinimide
N-(E-Maleimidocaproyloxy)sulphosuccinimide

Primary amine reactive -Sulphhydryl reactive Water soluble


Ref: Kitagawa,T.et al(1981) Chem Pharm.Bull. 29, 1130Ref: Keller,O.and Rudinger, J.Helv.Chem.Acta, 58, 531

Formel: C₁₄H₁₆N₂O₉S
Molecular Weight: 388,35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01310272
CAS-Nummer: 103848-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR