6-Maleinimido-capronsäure-N-succinimidylester (EMCS)

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dioxopyrrolidin-1-yl-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate

BICL222-250MG
APOSBICL222-250MGEA 21.9 EUR
APOSBICL222-250MG
6-Maleinimido-capronsäure-N-succinimidylester (EMCS)
6-Maleinimido-capronsäure-N-succinimidylester (EMCS)

Primary amine reactive -Sulphhydryl reactive.

Formel: C₁₄H₁₆N₂O₆
Molecular Weight: 308.29 g/mol
Schmelzpunkt: 70…73 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00043043
CAS-Nummer: 55750-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR