PMPI (4-Maleimidophenyl isocyanate)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
PMPI
BICL223-50MG
APOSBICL223-50MGEA
203
EUR
APOSBICL223-50MG
PMPI (4-Maleimidophenyl isocyanate)
PMPI (4-Maleimidophenyl isocyanate)
When melted, may contain small amount of sediment Hydroxyl reactive -Thiol reactive
Ref: Palumbo, P.S.et al(1993) Bioconjugate Chem. 4, 212-218
Formel:
C₁₁H₆N₂O₃ Molecular Weight: 214,177 g/mol Schmelzpunkt: 120…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00270900 CAS-Nummer: 123457-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...