3-(2-Pyridyldithio)propionic acid hydrazine hydrochloride
Lieferant: Apollo Scientific
BICL225-50MG
APOSBICL225-50MGEA 219 EUR
APOSBICL225-50MG
3-(2-Pyridyldithio)propionic acid hydrazine hydrochloride
3-(2-Pyridyldithio)propionic acid hydrazine hydrochloride
Sulphhydryl reactive -Carbohydrate reactive Cleavable