Maleimidopropionic acid hydrazide hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: MPH, 3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanehydrazide hydrochloride

BICL230-50MG
APOSBICL230-50MGEA 109 EUR
APOSBICL230-50MG
Maleimidopropionic acid hydrazide hydrochloride
Maleimidopropionic acid hydrazide hydrochloride
Formel: C₇H₉N₃O₃·ClH
Molecular Weight: 219,626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00467760
CAS-Nummer: 293298-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR