Dimethyl adipimidate dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Adipimidic Acid Dimethyl Ester Dihydrochloride

BICL300-10G
APOSBICL300-10GEA 79.9 EUR
APOSBICL300-10G
Dimethyl adipimidate dihydrochloride
Dimethyl adipimidate dihydrochloride
Formel: C₈H₁₆N₂O₂
Molecular Weight: 245,14 g/mol
Schmelzpunkt: 215…220 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012573
CAS-Nummer: 14620-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR