Benzoic acid-acyl-β-D-glucuronide ≥98%
Lieferant: Apollo Scientific
Synonyme:
E210-acyl-β-D-glucuronide, Benzoyl-acyl-β-D-glucuronide
BICL4036-1MG
BICL4036-5MG
BICL4036-2MG
APOSBICL4036-1MGEA
393
EUR
APOSBICL4036-1MG
APOSBICL4036-5MG
APOSBICL4036-2MG
Benzoic acid-acyl-β-D-glucuronide ≥98%
Benzoic acid-acyl-β-D-glucuronide
Lagertemperatur:
Raumtemperatur |
CAS-Nummer:
19237-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...