Benzoic acid-acyl-β-D-glucuronide ≥98%

Lieferant: Apollo Scientific

Synonyme: E210-acyl-β-D-glucuronide, Benzoyl-acyl-β-D-glucuronide

BICL4036-1MG BICL4036-5MG BICL4036-2MG
APOSBICL4036-1MGEA 393 EUR
APOSBICL4036-1MG APOSBICL4036-5MG APOSBICL4036-2MG
Benzoic acid-acyl-β-D-glucuronide ≥98%
Benzoic acid-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 19237-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR