Curcumin-4-O-β-D-glucuronide ≥85%

Lieferant: Apollo Scientific
Achtung

BICL4054-1MG
APOSBICL4054-1MGEA 207 EUR
APOSBICL4054-1MG
Curcumin-4-O-β-D-glucuronide ≥85%
Curcumin-4-O-β-D-glucuronide
Formel: C₂₇H₂₈O₁₂
Molecular Weight: 544,504 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28963761
CAS-Nummer: 227466-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR