2,3:4,5-Di-O-isopropylidene-D-arabitol 99% min

Lieferant: Apollo Scientific

BICL4060-1G BICL4060-5G BICL4060-2G
APOSBICL4060-1GEA 294 EUR
APOSBICL4060-1G APOSBICL4060-5G APOSBICL4060-2G
2,3:4,5-Di-O-isopropylidene-D-arabitol 99% min
2,3:4,5-Di-O-isopropylidene-D-arabitol
Formel: C₁₁H₂₀O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00063387
CAS-Nummer: 19139-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR