3,4:5,6-Di-O-isopropylidene-D-glucitol ≥99%

Lieferant: Apollo Scientific

BICL4064-1G BICL4064-5G BICL4064-2G
APOSBICL4064-1GEA 336 EUR
APOSBICL4064-1G APOSBICL4064-5G APOSBICL4064-2G
3,4:5,6-Di-O-isopropylidene-D-glucitol ≥99%
3,4:5,6-Di-O-isopropylidene-D-glucitol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 58846-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR