Mycophenolic acid-O-β-D-glucuronide ≥97%

Lieferant: Apollo Scientific

BICL4131-1MG BICL4131-5MG BICL4131-2MG
APOSBICL4131-1MGEA 253 EUR
APOSBICL4131-1MG APOSBICL4131-5MG APOSBICL4131-2MG
Mycophenolic acid-O-β-D-glucuronide ≥97%
Mycophenolic acid-O-β-D-glucuronide
Formel: C₂₃H₂₈O₁₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09878308
CAS-Nummer: 31528-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR