Mycophenolic acid acyl-β-D-glucuronide ≥90%

Lieferant: Apollo Scientific
Achtung

BICL4132-1MG BICL4132-5MG BICL4132-2MG
APOSBICL4132-1MGEA 393 EUR
APOSBICL4132-1MG APOSBICL4132-5MG APOSBICL4132-2MG
Mycophenolic acid acyl-β-D-glucuronide ≥90%
Mycophenolic acid acyl-β-D-glucuronide
Formel: C₂₃H₂₈O₁₂
Molecular Weight: 496.461 g/mol
Siedepunkt: 805 °C
Schmelzpunkt: 90 °C
Dichte: 1.5 g/cm³ (25 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD11977846
CAS-Nummer: 99043-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR