Methyl 2,3,4-tri-O-acetyl-1-O-trichloroacetimidoyl-α-D-glucopyranosyluronate ≥99%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,3S,4S,5R,6R)-2-(Methoxycarbonyl)-6-(2,2,2-trichloro-1-iminoethoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate, 2,3,4-Tri-O-acetyl-α-D-glucuronic acid methyl ester trichloroacetimidate

BICL4140-100MG
APOSBICL4140-100MGEA 119 EUR
APOSBICL4140-100MG
Methyl 2,3,4-tri-O-acetyl-1-O-trichloroacetimidoyl-α-D-glucopyranosyluronate ≥99%
Methyl 2,3,4-tri-O-acetyl-1-O-trichloroacetimidoyl-α-D-glucopyranosyluronate
Formel: C₁₅H₁₈Cl₃NO₁₀
Molecular Weight: 478.6600 g/mol
Schmelzpunkt: 125…126 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD02094293
CAS-Nummer: 92420-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR