Salicylic acid-acyl-β-D-glucuronide ≥98%

Lieferant: Apollo Scientific

BICL4196-1MG BICL4196-5MG BICL4196-2MG
APOSBICL4196-1MGEA 0 EUR
APOSBICL4196-1MG APOSBICL4196-5MG APOSBICL4196-2MG
Salicylic acid-acyl-β-D-glucuronide ≥98%
Salicylic acid-acyl-β-D-glucuronide
Lagertemperatur: Raumtemperatur
CAS-Nummer: 29315-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR