1,2:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
BICL4236-10G
BICL4236-2G
BICL4236-5G
APOSBICL4236-10GEA
978
EUR
APOSBICL4236-10G
APOSBICL4236-2G
APOSBICL4236-5G
1,2:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98%
1,2:4,5-Di-O-isopropylidene-β-D-fructopyranose
Formel:
C₁₂H₂₀O₆ Schmelzpunkt: 118…119 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00067639 CAS-Nummer: 25018-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...