Cyanidin-3-O-galactoside chloride 95% min

Lieferant: Apollo Scientific

Synonyme: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride

BICL4237-5MG BICL4237-1MG
APOSBICL4237-5MGEA 163 EUR
APOSBICL4237-5MG APOSBICL4237-1MG
Cyanidin-3-O-galactoside chloride 95% min
Cyanidin-3-O-galactoside chloride
Formel: C₂₁H₂₁O₁₁Cl
Molecular Weight: 484.85 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270387
CAS-Nummer: 27661-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR