Cholesterol-3-sulphate sodium salt 95% min

Lieferant: Apollo Scientific

BICL4238-1G BICL4238-250MG
APOSBICL4238-1GEA 0 EUR
APOSBICL4238-1G APOSBICL4238-250MG
Cholesterol-3-sulphate sodium salt 95% min
Cholesterol-3-sulphate sodium salt
Formel: C₂₇H₄₅O₄S·Na
Lagertemperatur: Raumtemperatur
MDL: MFCD00079091
CAS-Nummer: 2864-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR