5-Chloro-8-hydroxyquinoline sulphate ≥95%
Lieferant: Apollo Scientific
|
Achtung
|
BICL4277-10MG
BICL4277-25MG
BICL4277-5MG
APOSBICL4277-10MGEA
699
EUR
APOSBICL4277-10MG
APOSBICL4277-25MG
APOSBICL4277-5MG
5-Chloro-8-hydroxyquinoline sulphate ≥95%
5-Chloro-8-hydroxyquinoline sulphate
Schmelzpunkt:
186…188 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD28122834 CAS-Nummer: 15164-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...