Dihydroartemisinin O-β-D-glucuronide ≥95%

Lieferant: Apollo Scientific

BICL4328-1MG BICL4328-2MG
APOSBICL4328-1MGEA 434 EUR
APOSBICL4328-1MG APOSBICL4328-2MG
Dihydroartemisinin O-β-D-glucuronide ≥95%
Dihydroartemisinin O-β-D-glucuronide
Lagertemperatur: Raumtemperatur
MDL: MFCD08063733
CAS-Nummer: 198976-06-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR