3,4,6-Tri-O-acetyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%

Lieferant: Apollo Scientific

Synonyme: 3,4,6-Tri-O-acetyl-β-D-mannopyranose 1,2-(methyl orthoacetate), (3aS,5R,6R,7S,7aS)-5-(Acetoxymethyl)-2-methoxy-2-methyltetrahydro-5H-[1,3]dioxolo[4,5-b]pyran-6,7-diyl diacetate

BICL4374-1G
APOSBICL4374-1GEA 840 EUR
APOSBICL4374-1G
3,4,6-Tri-O-acetyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%
3,4,6-Tri-O-acetyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose
Formel: C₁₅H₂₂O₁₀
Molecular Weight: 362.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00211213
CAS-Nummer: 4435-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR