2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside 98%

Lieferant: Apollo Scientific

Synonyme: (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2-amino-6-chloro-9H-purin-9-yl)tetrahydrofuran-3,4-diyl diacetate, 2-Amino-6-chloro-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine

BICL5013-5G
APOSBICL5013-5GEA 184 EUR
APOSBICL5013-5G
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside 98%
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine riboside
Formel: C₁₆H₁₈ClN₅O₇
Molecular Weight: 427.80 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425473
CAS-Nummer: 16321-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR